Learn More
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 100-42-5, 98-29-3 |
---|---|
Molecular Formula | C8H8 |
Molecular Weight (g/mol) | 104.15 |
MDL Number | MFCD00008612,MFCD00084450 |
InChI Key | PPBRXRYQALVLMV-UHFFFAOYSA-N |
Synonym | ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene |
PubChem CID | 7501 |
ChEBI | CHEBI:27452 |
IUPAC Name | styrene |
SMILES | C=CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031380
|
Thermo Scientific Acros
132790100 |
10 mL | Glass Bottle |
N/A
|
N/A | ||||
10021380
|
Thermo Scientific Acros
132790010 |
1 L | Glass Bottle |
N/A
|
N/A | ||||
10103190
|
Thermo Scientific Acros
132790025 |
2.5 L | Glass Bottle |
N/A
|
N/A | ||||
10398093
|
Thermo Scientific Acros
132790250 |
25 L | Metal drum |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-42-5, 98-29-3 | |
104.15 | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
7501 | |
styrene |
C8H8 | |
MFCD00008612,MFCD00084450 | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
CHEBI:27452 | |
C=CC1=CC=CC=C1 |
Specifications
100-42-5, 98-29-3 | |
0.9060g/mL | |
31°C | |
98.5% min. (GC) | |
C6H5CH=CH2 | |
MFCD00008612,MFCD00084450 | |
15, 8990 | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
styrene | |
104.15 | |
CHEBI:27452 | |
104.15 | |
99% | |
Liquid |
-31.0°C | |
145.0°C to 146.0°C | |
Authentic | |
C8H8 | |
1.5460 to 1.5480 | |
05, 474 | |
0.906 | |
Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone,cs2,ccl4,dissolves organic substances and polymers | |
C=CC1=CC=CC=C1 | |
10 to 20ppm p-tert-butylcatechol | |
7501 | |
0.695 mPa.s (25°C) | |
7mbar at 20°C | |
Extra Pure | |
Styrene, Stabilized |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
May be fatal if swallowed and enters airways.<br/
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/prote
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-851-5
- Styrene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H372-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only