Learn More
Thermo Scientific™ Subcellular Protein Fractionation Kit for Cultured Cells
Enrich for cultured cell-specific cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal proteins in less than 3 hours.
Brand: Thermo Scientific™ 78840
517.64 GBP valid until 2025-12-31
Use promo code "24353" to get your promotional price.
Description
Perform segregation and enrichment of proteins from five different cellular compartments with Thermo Scientific™ Subcellular Protein Fractionation Kit for Cultured Cells.
The kit enables the stepwise separation and extraction of cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal proteins from mammalian cultured cells. Extracts obtained with the Subcellular Protein Fractionation Kit for Cultured Cells are compatible with a variety of downstream applications including Western blotting, protein assays, electrophoretic mobility shift assays and reporter-gene and enzyme-activity assays. Fractionation of subcellular proteins enables protein localization assessment and protein enrichment from specific cellular compartments. The Thermo Scientific Subcellular Protein Fractionation Kit includes a combination of reagents for stepwise lysis of cells into functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than 3 hours. Extracts from each subcellular compartment generally have less than 15% contamination between fractions, which is sufficient purity for most experiments studying protein localization and redistribution.Highlights:
Concise – extract functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than three hours
Convenient – perform a simple procedure without using ultracentrifugation over gradients
Complete – obtain cytoplasmic, membrane, soluble nuclear, chromatin-bound and cytoskeletal protein fractions from a single kit
Cultured Cells – developed and optimized for use with cultured mammalian cells; for tissue samples use the Subcellular Fractionation Kit for Tissues, Part No. 87790
Compatible – use with a variety of downstream applications including Western blotting, protein assays, electrophoretic mobility shift assays and reporter-gene and enzyme-activity assays
Small-scale – kit provides sufficient reagents to fractionate 50 cell pellets of 20μL each, equivalent to a total of 2g of cell paste or tissue
Includes:
Four extraction buffers, micrococcal nuclease enzyme, calcium chloride solution and protease inhibitor cocktail
Recommended for:
Extraction and enrichment of proteins from different cellular compartments; Separation and profiling of protein from the cytoplasm, nucleus, membrane and cytoskeleton from the same cell sample; Studying protein translocation

Specifications
Subcellular Protein Fractionation Kit | |
Liquid | |
Cell Lysis Buffer, Detergent Solution, Subcellular Fractionation | |
1 Kit |
Upon receipt store kit at -20°C, or store individual components as indicated above. Kit is shipped with dry ice. | |
Lysis Buffer | |
50 Samples |
Safety and Handling
- Subcellular Protein Fractionation Kit for Cultured Cells
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 4-(2-aminoethyl)-Benzenesulfonyl fluoride hydrochloride
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.