Learn More
Suberoyl chloride, 97%, Thermo Scientific Chemicals
£61.41 - £188.94
Chemical Identifiers
CAS | 10027-07-3 |
---|---|
Molecular Formula | C8H12Cl2O2 |
Molecular Weight (g/mol) | 211.082 |
MDL Number | MFCD00000764 |
InChI Key | PUIBKAHUQOOLSW-UHFFFAOYSA-N |
Synonym | suberoyl chloride, suberoyl dichloride, suberic acid dichloride, suberyl chloride, suberyl dichloride, acmc-1bonz, octanedioic acid dichloride |
PubChem CID | 534653 |
IUPAC Name | octanedioyl dichloride |
SMILES | C(CCCC(=O)Cl)CCC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430428
|
Thermo Scientific Alfa Aesar
B24580.06 |
5 g |
£61.41
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440428
|
Thermo Scientific Alfa Aesar
B24580.14 |
25 g |
£188.94
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSuberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.
Solubility
Reacts with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
Chemical Identifiers
10027-07-3 | |
211.082 | |
PUIBKAHUQOOLSW-UHFFFAOYSA-N | |
534653 | |
C(CCCC(=O)Cl)CCC(=O)Cl |
C8H12Cl2O2 | |
MFCD00000764 | |
suberoyl chloride, suberoyl dichloride, suberic acid dichloride, suberyl chloride, suberyl dichloride, acmc-1bonz, octanedioic acid dichloride | |
octanedioyl dichloride |
Specifications
10027-07-3 | |
162°C to 163°C (15 mmHg) | |
C8H12Cl2O2 | |
1.468 | |
UN3265 | |
suberoyl chloride, suberoyl dichloride, suberic acid dichloride, suberyl chloride, suberyl dichloride, acmc-1bonz, octanedioic acid dichloride | |
PUIBKAHUQOOLSW-UHFFFAOYSA-N | |
octanedioyl dichloride | |
534653 | |
97% |
1.172 | |
>110°C (230°F) | |
ClOC(CH2)6COCl | |
MFCD00000764 | |
Moisture sensitive | |
Reacts with water. | |
C(CCCC(=O)Cl)CCC(=O)Cl | |
211.082 | |
211.09 | |
Suberoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only