Learn More
DiscoverX Succinic acid disodium salt
Succinic acid disodium salt is an agonist for GPR91 (SUCNR1). Provided as 10mg of dry powder.
Brand:  DiscoverX 92-1193
Encompass
Product Code. 17322519
  • £111.09 / 10mg
Explore more special offers
Description
Description
Succinic acid is a dicarboxylic acid which takes the form of succinate in living organisms. Succinate is produced in inflammatory conditions, may be associated with rheumatoid arthritis, and its accumulation is associated with tumorigenesis. It acts through the succinate receptor, SUCNR1 (or GPR91), leading to enhanced IL-1β production. Provided as 10mg of dry powder.
Specifications
Specifications
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DiscoverX Succinic acid disodium salt