Learn More
Thermo Scientific Chemicals Sucralose, 98%
£52.46 - £174.87
Chemical Identifiers
CAS | 56038-13-2 |
---|---|
Molecular Formula | C12H19Cl3O8 |
Molecular Weight (g/mol) | 397.63 |
MDL Number | MFCD03648615 |
InChI Key | BAQAVOSOZGMPRM-QBMZZYIRSA-N |
Synonym | sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit |
PubChem CID | 71485 |
ChEBI | CHEBI:32159 |
IUPAC Name | (2R,3R,4R,5R,6R)-2-[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy-5-chloro-6-(hydroxymethyl)oxane-3,4-diol |
SMILES | OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15418209
|
Thermo Scientific Chemicals
J66736.18 |
50 g |
£52.46
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15428209
|
Thermo Scientific Chemicals
J66736.22 |
100 g |
£89.24
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15438209
|
Thermo Scientific Chemicals
J66736.30 |
250 g |
£174.87
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.
Solubility
Soluble in Water.
Notes
Research use only: Not intended for food, cosmetic, or diagnostic and therapeutic use in animals nor humans.
Chemical Identifiers
56038-13-2 | |
397.63 | |
BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
71485 | |
(2R,3R,4R,5R,6R)-2-[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy-5-chloro-6-(hydroxymethyl)oxane-3,4-diol |
C12H19Cl3O8 | |
MFCD03648615 | |
sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit | |
CHEBI:32159 | |
OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl |
Specifications
56038-13-2 | |
White | |
MFCD03648615 | |
14,8880 | |
Soluble in Water. | |
OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl | |
397.63 | |
CHEBI:32159 | |
98% | |
Sucralose |
130°C (decomposition) | |
C12H19Cl3O8 | |
3654410 | |
sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit | |
BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
(2R,3R,4R,5R,6R)-2-[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy-5-chloro-6-(hydroxymethyl)oxane-3,4-diol | |
71485 | |
397.63 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 259-952-2
missing translation for 'rtecsNumber' : LW5440140
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.