missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Sucralose, 98% Product Code.: 15418209

Thermo Scientific Chemicals Sucralose, 98%

Product Code. 15418209
50 g
Click to view available options
Quantity:
50 g
100 g
250 g
This item is not returnable. View return policy

Product Code. 15418209

Brand: Thermo Scientific Chemicals J66736.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.

Solubility
Soluble in Water.

Notes
Research use only: Not intended for food, cosmetic, or diagnostic and therapeutic use in animals nor humans.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56038-13-2
Molecular Formula C12H19Cl3O8
Molecular Weight (g/mol) 397.63
MDL Number MFCD03648615
InChI Key BAQAVOSOZGMPRM-QBMZZYIRSA-N
Synonym sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvitShow MoreShow Less
PubChem CID 71485
ChEBI CHEBI:32159
IUPAC Name (2R,3R,4R,5R,6R)-2-[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy-5-chloro-6-(hydroxymethyl)oxane-3,4-diol
SMILES OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

Specifications

Melting Point 130°C (decomposition)
Color White
Quantity 50 g
Beilstein 3654410
Merck Index 14,8880
Solubility Information Soluble in Water.
Formula Weight 397.63
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sucralose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS14K020 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog NumberJ66736.30,J66736.22,J66736.18
Lot NumberN07I043 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ66736.30,J66736.22,J66736.18
Lot NumberY13F055 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66736.30,J66736.22,J66736.18
Lot NumberZ26E077 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66736.30,J66736.22,J66736.18
Lot NumberP20B001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66736.30,J66736.22,J66736.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sucralose, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.