Learn More
Sudan orange G, pure, Thermo Scientific Chemicals
£18.60 - £824.00
Chemical Identifiers
CAS | 2051-85-6 |
---|---|
Molecular Formula | C12H10N2O2 |
Molecular Weight (g/mol) | 214.22 |
MDL Number | MFCD00002275 |
InChI Key | PULNESMFDLBKAZ-SDNWHVSQSA-N |
Synonym | C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1 |
PubChem CID | 5921655 |
IUPAC Name | (4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one |
SMILES | OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10736822
|
Thermo Scientific Acros
190170050 |
5 g | Glass bottle |
£18.60
5g |
In Stock
Log in to see stock. |
||||
10481582
|
Thermo Scientific Acros
190170250 |
25 g | Glass bottle |
£57.90
25g |
In Stock
Log in to see stock. |
||||
10350494
|
Thermo Scientific Acros
190171000 |
100 g | Glass bottle |
£164.00
100g |
In Stock
Log in to see stock. |
||||
10301394
|
Thermo Scientific Acros
190170010 |
1 kg | Glass bottle |
£824.00
1kg |
Estimated Shipment: 24-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2051-85-6 | |
214.22 | |
PULNESMFDLBKAZ-SDNWHVSQSA-N | |
5921655 | |
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1 |
C12H10N2O2 | |
MFCD00002275 | |
C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1 | |
(4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one |
Specifications
2051-85-6 | |
MFCD00002275 | |
PULNESMFDLBKAZ-SDNWHVSQSA-N | |
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1 | |
214.22 | |
214.22 | |
Complies | |
Powder |
C12H10N2O2 | |
C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1 | |
Authentic | |
(4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one | |
5921655 | |
Extra Pure | |
Orange to Red | |
Sudan orange G, Pure |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 218-131-9
missing translation for 'rtecsNumber' : CZ9027500
missing translation for 'tsca' : TSCA
- Sudan Orange G
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only