missing translation for 'onlineSavingsMsg'
Learn More
Sulfamic acid, 99%, Thermo Scientific Chemicals Product Code.: 10432031

Sulfamic acid, 99%, Thermo Scientific Chemicals

Product Code. 10432031
1 kg, Plastic Bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10432031

Brand: Thermo Scientific Acros 222070010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.09
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acidShow MoreShow Less
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Color White
Melting Point 200.0°C to 208.0°C
Packaging Plastic Bottle
Assay Percent Range 99%
Quantity 1 kg
Linear Formula H2NSO3H
Physical Form Crystals or Crystalline Powder
Fieser 11117
Merck Index 159053
Solubility Information Solubility in water: 146.8g/L (20°C). Other solubilities: sparingly soluble in alcohol and methanol,slightly soluble in acetone,insoluble in ether
Percent Purity 99%
Chemical Name or Material Sulfamic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465494 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number222070025,222072500,222070100,222070010
Lot NumberA0457886 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number222070025,222072500,222070100,222070010
Lot NumberA0452907 Certificate TypeCertificate of Analysis Date05/05/2023 Catalog Number222070025,222072500,222070100,222070010
Lot NumberA0448750 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number222070025,222072500,222070100,222070010
Lot NumberA0399612 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number222070025,222072500,222070100,222070010
compliance-icons
Product Identifier
  • Sulfamic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfamic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.