missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Melting Point | 200.0°C to 208.0°C |
Packaging | Plastic Bottle |
Assay Percent Range | 99% |
Quantity | 2.5 kg |
Linear Formula | H2NSO3H |
Physical Form | Crystals or Crystalline Powder |
Fieser | 11117 |
Merck Index | 159053 |
Solubility Information | Solubility in water: 146.8g/L (20°C). Other solubilities: sparingly soluble in alcohol and methanol,slightly soluble in acetone,insoluble in ether |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465494 | Certificate of Analysis | 19/08/2024 | 222070025,222072500,222070100,222070010 |
A0457886 | Certificate of Analysis | 27/11/2023 | 222070025,222072500,222070100,222070010 |
A0452907 | Certificate of Analysis | 05/05/2023 | 222070025,222072500,222070100,222070010 |
A0448750 | Certificate of Analysis | 05/01/2023 | 222070025,222072500,222070100,222070010 |
A0399612 | Certificate of Analysis | 16/12/2022 | 222070025,222072500,222070100,222070010 |
Safety and Handling

Product Identifier
- Sulfamic acid
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : Sulfamic acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sulfamic acid, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction