Learn More
Sulfur Oil Standard in #2 Diesel Fuel, 1000μg/g, Set, SPEX CertiPrep™
Sulfur Oil Standard Set in #2 Diesel Fuel. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Brand: SPEX Certiprep SDFS-SET
Code : NEW
Additional Details : CAS Number : 8002-05-9 Weight : 0.09000kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Sulfur Oil Standard Set
- For use with ultra low sulfur standards
- Includes sulfur standards in #2 Diesel Oil in concentrations of 5, 10, 15, 20, 25, 50 and 100 μg/g
- Also includes Sulfur Blank
- 100 mL volume fo
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Sulfur Oil Standards in #2 Diesel Fuel | |
For Diesel Fuel Analysis | |
8 x 125 mL | |
Triple-leached LDPE bottle |
Product intended for laboratory use only