missing translation for 'onlineSavingsMsg'
Learn More
Sulfur pieces, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals Product Code.: 11375778

Sulfur pieces, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Product Code. 11375778
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11375778

Brand: Thermo Scientific Alfa Aesar 010343.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Color Yellow
Physical Form Pieces
Quantity 100 g
Assay Percent Range 99.9995% (Metals basis)
Ignition Point 248°C (478°F)
Odor Weak
Appearance Pale yellow pieces
Chemical Name or Material Sulphur

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ20K075 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number010343.36,010343.14,010343.22
Lot NumberS21K009 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number010343.14,010343.36,010343.22
Lot NumberQ04K006 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number010343.14,010343.36,010343.22
Lot NumberY09I007 Certificate TypeCertificate of Analysis Date22/11/2022 Catalog Number010343.14,010343.36,010343.22
Lot NumberU12G018 Certificate TypeCertificate of Analysis Date28/04/2022 Catalog Number010343.14,010343.36,010343.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfur pieces, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.