Learn More
Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%, Thermo Scientific Chemicals
£17.30 - £347.00
Chemical Identifiers
CAS | 26412-87-3 |
---|---|
Molecular Formula | C5H5NO3S |
Molecular Weight (g/mol) | 159.16 |
MDL Number | MFCD00012437 |
InChI Key | UDYFLDICVHJSOY-UHFFFAOYSA-N |
Synonym | pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide |
PubChem CID | 168533 |
SMILES | O=S(=O)=O.C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11423960
|
Thermo Scientific Alfa Aesar
A12202.09 |
10 g |
£17.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474863
|
Thermo Scientific Alfa Aesar
A12202.18 |
50 g |
£33.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484863
|
Thermo Scientific Alfa Aesar
A12202.30 |
250 g |
£111.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464863
|
Thermo Scientific Alfa Aesar
A12202.0B |
1000 g |
£347.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.
Solubility
Soluble in polar organic solvents.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
26412-87-3 | |
159.16 | |
UDYFLDICVHJSOY-UHFFFAOYSA-N | |
168533 |
C5H5NO3S | |
MFCD00012437 | |
pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide | |
O=S(=O)=O.C1=CC=NC=C1 |
Specifications
26412-87-3 | |
C5H5NO3S | |
UN3261 | |
Moisture sensitive | |
Soluble in polar organic solvents. | |
O=S(=O)=O.C1=CC=NC=C1 | |
159.16 | |
159.16 | |
Sulfur trioxide-pyridine complex, Active SO3 ≈48 to 50% |
∼155°C to 165°C | |
MFCD00012437 | |
3704116 | |
pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide | |
UDYFLDICVHJSOY-UHFFFAOYSA-N | |
pyridine;sulfur trioxide | |
168533 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 247-683-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only