Learn More
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 124240010
37.64 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Sulfuric acid | |
1N (0.5M) Standard Solution | |
0.998 to 1.002N (20°C) | |
Plastic bottle | |
H2O4S | |
Liquid | |
01,470; 05,633; 06,558; 07,347; 09,441 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
Pure |
Colorless | |
7664-93-9, 7732-18-5 | |
1.05g/mL | |
1 L | |
H2SO4 | |
MFCD00064589 | |
15, 9104 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.07 |
Safety and Handling
- Sulfuric acid, 1N standard solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : Sulfuric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only