missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation SunFire C18 Column Method Validation Kit

This kit consists of three Sunfire C18 columns packed with three different batches of sorbent.

Brand:  Waters Corporation 186002675

Encompass

 View more versions of this product

Product Code. 13250450

  • £2880.00 / Pack of 3
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes three Sunfire C18 columns packed with three different batches of sorbent.
Specifications

Specifications

4.6 mm
6000 psi (415 Bar)
3.5 μm
3/Pk.
Yes
100 mm
Spherical
100 Å
L1
Method Validation Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers