missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Erecta™ Chrome Wire Shelf, Brite Finish

Design your storage to suit your lab's needs. Metro Super Erecta Chrome Wire Shelf in Brite finish is easily adjustable, offering multiple storage configurations when used with Metro Super Erecta shelving systems.

Brand:  Metro™ 1836BR

 View more versions of this product

Product Code. 15966591

  • £85.40 / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore más ofertas especiales
Añadir a la cesta

Includes: Plastic sleeves and wedges.
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

  • Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
  • Economical chromate finish for dry, low humidity environment
  • Lacquer coated finish provides the look of chrome

Especificaciones

Especificaciones

36 x 18 in. (91.4 x 45.7 cm)
Metro™ Super Erecta™ shelving system
36 in.
800 lb.
18 in.
Chrome
Brite
Plastic sleeves and wedges
91.4 cm
363 kg
45.7 cm
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales