Learn More
Metro™ Super Erecta™ Chrome Wire Shelf, Brite Finish
Design your storage to suit your lab's needs. Metro Super Erecta Chrome Wire Shelf in Brite finish is easily adjustable, offering multiple storage configurations when used with Metro Super Erecta shelving systems.
£66.45 - £154.00
Specifications
Finish | Brite |
---|---|
For Use With (Equipment) | Metro™ Super Erecta™ shelving system |
Includes | Plastic sleeves and wedges |
Material | Chrome |
Product Code | Brand | Load Bearing Capacity (English) | Load Bearing Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity (English) | Load Bearing Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
15916581
|
Metro™
1460BR |
600 lb. | 272 kg | 60 x 14 in. (152.4 x 35.5 cm) |
£94.85
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15906621
|
Metro™
2130BR |
800 lb. | 363 kg | 30 x 21 in. (76 x 53 cm) |
£91.50
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15946651
|
Metro™
2430BR |
800 lb. | 363 kg | 30 x 24 in. (76 x 61 cm) |
£94.85
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15976561
|
Metro™
1424BR |
800 lb. | 363 kg | 24 x 14 in. (61 x 35.5 cm) |
£66.45
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15916601
|
Metro™
1842BR |
800 lb. | 363 kg | 42 x 18 in. (106.6 x 45.7 cm) |
£93.25
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15936611
|
Metro™
1872BR |
600 lb. | 272 kg | 72 x 18 in. (183 x 45.7 cm) |
£132.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15976601
|
Metro™
1854BR |
600 lb. | 272 kg | 54 x 18 in. (137 x 45.7 cm) |
£113.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15936591
|
Metro™
1830BR |
800 lb. | 363 kg | 30 x 18 in. (76 x 45.7 cm) |
£80.20
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15996601
|
Metro™
1860BR |
600 lb. | 272 kg | 60 x 18 in. (152.4 x 45.7 cm) |
£113.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15926671
|
Metro™
2454BR |
600 lb. | 272 kg | 54 x 24 in. (137 x 61 cm) |
£143.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15966591
|
Metro™
1836BR |
800 lb. | 363 kg | 36 x 18 in. (91.4 x 45.7 cm) |
£82.45
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
|||
15996621
|
Metro™
2142BR |
800 lb. | 363 kg | 42 x 21 in. (106.6 x 53 cm) |
£107.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15946661
|
Metro™
2442BR |
800 lb. | 363 kg | 42 x 24 in. (106.6 x 61 cm) |
£117.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15956571
|
Metro™
1442BR |
800 lb. | 363 kg | 42 x 14 in. (106.6 x 35.5 cm) |
£81.90
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15926641
|
Metro™
2160BR |
600 lb. | 272 kg | 60 x 21 in. (152.4 x 53 cm) |
£127.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15926571
|
Metro™
1436BR |
800 lb. | 363 kg | 36 x 14 in. (91.4 x 35.5 cm) |
£71.85
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15906651
|
Metro™
2424BR |
800 lb. | 363 kg | 24 x 24 in. (61 x 61 cm) |
£92.15
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15936601
|
Metro™
1848BR |
800 lb. | 363 kg | 48 x 18 in. (122 x 45.7 cm) |
£93.25
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15956611
|
Metro™
2124BR |
800 lb. | 363 kg | 24 x 21 in. (61 x 53 cm) |
£89.40
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15956621
|
Metro™
2136BR |
800 lb. | 363 kg | 36 x 21 in. (91.4 x 53 cm) |
£91.50
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15966581
|
Metro™
1472BR |
600 lb. | 272 kg | 72 x 14 in. (183 x 35.5 cm) |
£113.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15996631
|
Metro™
2154BR |
600 lb. | 272 kg | 54 x 21 in. (137 x 53 cm) |
£127.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15946631
|
Metro™
2148BR |
800 lb. | 363 kg | 48 x 21 in. (123 x 53 cm) |
£107.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15966641
|
Metro™
2172BR |
600 lb. | 272 kg | 72 x 21 in. (183 x 53 cm) |
£154.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
15986651
|
Metro™
2436BR |
800 lb. | 363 kg | 36 x 24 in. (91.4 x 61 cm) |
£94.85
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
Description
- Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
- Economical chromate finish for dry, low humidity environment
- Lacquer coated finish provides the look of chrome
Specifications
Brite | |
Metro™ Super Erecta™ shelving system | |
Plastic sleeves and wedges | |
Chrome |