Learn More
Thermo Scientific™ SuperBlock™ Blocking Buffer
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
£62.25 - £746.00
Specifications
For Use With (Application) | ELISA, Western Blot |
---|---|
Product Line | SuperBlock™ |
Blocking Agent | Single purified protein |
Content And Storage | Upon receipt store at 4°C. Product is shipped at ambient temperature. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13434299
|
Thermo Scientific™
37580 |
SuperBlock (PBS) Blocking Buffer, 100 mL |
£62.25
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10076713
|
Thermo Scientific™
37515 |
SuperBlock (PBS) Blocking Buffer, 1 L |
£283.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11884524
|
Thermo Scientific™
37518 |
SuperBlock (PBS) Blocking Buffer, 5 L |
£746.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13444299
|
Thermo Scientific™
37581 |
SuperBlock (TBS) Blocking Buffer, 100 mL |
£62.25
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10016723
|
Thermo Scientific™
37535 |
SuperBlock (TBS) Blocking Buffer, 1 L |
£251.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10426144
|
Thermo Scientific™
37545 |
SuperBlock (TBS) Blocking Buffer, dry blend, 5 packs |
£226.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10519293
|
Thermo Scientific™
37516 |
SuperBlock T20 (PBS) Blocking Buffer, 1 L |
£267.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10668654
|
Thermo Scientific™
37536 |
SuperBlock T20 (TBS) Blocking Buffer, 1 L |
£267.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.
Features of SuperBlock Blocking Buffer include:• Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
• Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
• Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.
Specifications
ELISA, Western Blot | |
SuperBlock™ | |
Single purified protein | |
Upon receipt store at 4°C. Product is shipped at ambient temperature. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.