Learn More
Cytiva Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns
For size exclusion chromatography and analysis of proteins with molecular weights between 10,000 and 600,000. Cytiva Lifescience™ Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns are designed for small scale preparative purification.
Brand: Cytiva 28990944
2116.54 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : GH
Additional Details : CAS Number : 64-17-5 Weight : 0.35460kg
Description
- For small-scale (25 to 500μL) preparative purification and for analytical purposes, suited for antibodies
- Allows characterization of proteins, aggregate analysis of antibodies, size homogeneity analysis of membrane proteins, or protein-protein interactions
- Prepacked with Superdex 200 Increase media
- Composite of cross-linked high-flow agarose and dextran base matrix for high flow rates
- Smaller bead size of average 8.6μm for increased resolution
- Flow Rate: <1.80mL/min1
- Bed Volume: Approximately 24mL
- Column 1.D.: 10mm
- Columns are supplied with two fingertight connectors 1/16 in. male for connection to AKTA™ or other system
Specifications
25 to 500μL | |
Superdex Pre-packed Column | |
Small-scale (25 to 500μL) preparative purification and analysis | |
Cross-linked agarose and dextran |