Learn More
Cytiva Superdex™ 200 Increase 3.2/300 Prepacked Precision Column
Used for micro-scale (μg) preparative purification and for characterization and analysis of small volumes (4 to 50μL) of proteins. Cytiva Lifescience™ Superdex™ 200 Increase 3.2/300 Prepacked Precision Column desigened for size exclusion chromatography.
Brand: Cytiva 28990946
Code : GH
Additional Details : CAS Number : 64-17-5 Weight : 0.30000kg
Description
- Prepacked with Superdex 200 Increase media
- For proteins with molecular weights between 10,000 and 6,00,000 such as antibodies
- Composite of cross-linked high-flow agarose and dextran base matrix for high flow rates
- Used for preparative and analytical procedures
- Smaller bead size average 8.6μm for higher resolution
- Supplied with two fingertight 1/16 in. male connectors for connection
Specifications
<0.15 mL/min.1 | |
8.6 μm | |
Superdex Pre-packed Column | |
Borosilicate Glass |
Safety and Handling
- Superdex 200 Increase, 3.2/300
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.