Learn More
Cytiva Superdex™ 200 Increase 5/150 GL Prepacked Tricorn™ Columns
For size exclusion chromatography and analysis of proteins with molecular weights between 10,000 and 600,000. Cytiva Lifescience™ Superdex™ 200 Increase 5/150 GL Prepacked Tricorn™ Columns are designed for small scale preparative purification.
Brand: Cytiva 28990945
Code : GH
Additional Details : CAS Number : 64-17-5 Weight : 0.22560kg
Description
- For rapid purity checks and different experimental conditions screenings
- Prepacked with Superdex 200 Increase media
- Composite of cross-linked high-flow agarose and dextran base matrix for high flow rates
- Smaller bead size of average 8.6μm for increased resolution
- Flow Rate: <0.75mL/min1
- Bed Volume: Approximately 3mL
- Column 1.D.: 5mm
- Columns are supplied with two fingertight connectors 1/16 in. male for connection to AKTA™ or other system
Specifications
4 to 50μL | |
Superdex 200 Increase | |
Rapid purity checks and quick screenings | |
Cross-linked agarose and dextran |
Safety and Handling
- Superdex 200 Increase, 5/150 GL
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.