missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Superdex 30 Increase Precision 3.2/300 GL Prepacked SEC Column
Designed for high resolution in small-scale preparative purification and analysis for sample volumes up to 500μL
Brand: Cytiva 29219758
2205.91 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement for Superdex peptide columns
- Reduced run time with maintained resolution for time saving or increased throughput
- Higher resolution for improved protein purity and analysis data
- Higher lot-to-lot consistency for
Specifications
Each | |
∼9 μm1 | |
Superdex Pre-packed Column | |
High resolution, small-scale purification and analysis of small biomolecules with Mr ˜ 100 to ˜7000, such as peptides | |
3 to 12 | |
Superdex Increase |
0.075 mL/min1 | |
2.4mL | |
3.2mm | |
Column tube: Borosilicate Glass; Bed Support: Titatium | |
Frit | |
4°C to 30°C, 20% ethanol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Superdex 30 Increase 3.2/300
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction