missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Superdex 30 Increase Tricon™ 10/300 GL Prepacked SEC Column
Designed for high resolution in small-scale preparative purification and analysis for sample volumes up to 500μL
Brand: Cytiva 29219757
This item is not returnable.
View return policy
Description
- Replacement for Superdex peptide columns
- Reduced run time with maintained resolution for time saving or increased throughput
- Higher resolution for improved protein purity and analysis data
- Higher lot-to-lot consistency for
Specifications
Each | |
∼9 μm1 | |
Superdex Pre-packed Column | |
High resolution, small-scale purification and analysis of small biomolecules with Mr ˜ 100 to ˜7000, such as peptides | |
Column tube: Borosilicate Glass; Bed Support: Polyethylene (PE) filter | |
Filter | |
4°C to 30°C, 20% ethanol |
<1.2 mL/min.1 | |
24mL | |
10mm | |
279 to 309 mm | |
3 to 12 | |
Superdex Increase |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction