Learn More
Cytiva Superdex 30 PG size exclusion chromatography media prepacked into HiLoad 26/600 column
Superdex 30 prep grade (PG) size exclusion chromatography (gel filtration) media prepacked into a HiLoad 26/600 column is optimized for the separation of proteins and peptides, but can also be used with success to separate oligosaccharides.
Brand: Cytiva 28-9893-32
Code : GH
Additional Details : Weight : 1.90000kg
Includes: Dismantling tool, support screen, net ring (10μm), O-ring, stop plugs, and instructions.
Description
- High-resolution separation – with short run times and good recovery.
- High capacity – at high flow rates.
- Prepacked – for convenient ready-to-go use and reproducible results.
- Scalable protein separation – easily scale-up separations to production levels.
- Integrated workflows – 1/16” fittings for convenient connection with ÄKTA protein purification systems.
Recommended Storage
4 to 30°C, 20% Ethanol
Specifications
< 2.6 mL/min. | |
320 mL | |
26 mm | |
3 bar [0.3 MPa] (42 psi) | |
Dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs, and instructions. | |
4 to 30°C, 20% Ethanol |
34 μm | |
Superdex Pre-packed Column | |
Superdex 30 prep grade | |
3 to 12 | |
1 × 320mL |