missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Superdex™ 30 Prep Grade
Offers high resolution with short run times and good recovery
Brand: Cytiva 17-0905-01
This item is not returnable.
View return policy
Description
- Gel filtration medium
- For purifying proteins of molecular weight range up to 10000
- Average particle size: 34μm
- Matrix: composite of cross-linked agarose and dextran
- pH stability working range
- Available in 150mL bottle
Specifications
150 mL | |
24 to 44 μm | |
Solid Spherical Bead | |
70psi | |
Composite of cross-linked Agarose and Dextran | |
4 to 30°C, 0.2 M sodium acetate in 20% Ethanol |
10 to 50 cm/hr. | |
Superdex Media | |
Separation, polishing and product formulation of Peptides and small Proteins | |
3 to 12 | |
Peptides and small Proteins |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Superdex 30 prep grade, 150 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction