missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Superose™ 6 3.2/300 Prepacked Precision Column
Prepacked with Superose 6 media. Cytiva Superose™ 6 3.2/300 Prepacked Precision Column separate a broad fractional range for biomolecules with molecular weight between 5000 and 5,00,000.
Brand: Cytiva 29036226
Code : A0
Additional Details : Weight : 0.30000kg
Description
- Cross-linked agarose matrix
- Supplied with two fingertight 1/16 in. male connectors for connection
- With new hardware design for direct connection to ÄKTATM systems
Specifications
0.01 mL/min. to 0.10mL/min1 | |
11 to 15 μm | |
Borosilicate Glass | |
Superdex Pre-packed Column |