Learn More
Cytiva Superose™ 6 Increase Columns
High-resolution size-exclusion chromatography for studying large proteins and complexes. Cytiva Lifescience™ Superose™ 6 Increase Columns delivers high quality in size exclusion chromatography (SEC).
£2299.00 - £2420.00
Specifications
Material | Borosilicate glass |
---|---|
Ligand Type | Cross-linked agarose |
Max. Pressure | 435 psi |
pH Range | 3 to 12 |
Particle Size | 8.6μm |
Product Code | Brand | Diameter (Metric) Inner | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Height (Metric) | Price | Quantity & Availability | ||||
15377653
|
Cytiva
29091596 |
10 mm | 300 mm |
£2390.96
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15303234
|
Cytiva
29091598 |
3.2 mm | 300 mm |
£2420.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15383224
|
Cytiva
29091597 |
5 mm | 150 mm |
£2299.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Superose™ 6 Increase pre-packed columns deliver the quality and high resolution in size exclusion chromatography (SEC) — even for large proteins and aggregates.
Increased performance
- Higher resolution than traditional Superose 6 columns
- Wide fractionation range (Mr from 5,000 to 5,000,000)
- Withstands high-pH cleaning
Specifications
Borosilicate glass | |
435 psi | |
8.6μm | |
Gel Filtration Column |
Cross-linked agarose | |
3 to 12 | |
4° to 30° in 20% ethanol |
Safety and Handling
- Superose 6 Increase 10/300 GL
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.