Learn More
Invitrogen™ SuperScript™ IV RT-LAMP Master Mix
Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions.
Brand: Invitrogen™ A51801
157.60 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions. With this convenient master mix formulation, you can perform isothermal amplification from both RNA or DNA targets in a single tube format.
Benefits of SuperScript IV RT-LAMP Master Mix
• RNA and DNA target detection using Bst DNA polymerase for isothermal amplification in as little as five minutes
• Robust and specific cDNA synthesis using SuperScript IV Reverse Transcriptase
• Enhanced specificity and sensitivity with buffer formulation and RNaseOUT recombinant ribonuclease inhibitor
• Compatible with real-time and endpoint detection—includes SYTO 9 dye for real-time detection
• Flexible design provides several options for evaluating results, including real-time and endpoint detection methods
SuperScript IV RT-LAMP Master Mix includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance. To prevent RNA degradation by carry-over RNases, the master mix includes the potent, noncompetitive RNaseOUT Recombinant Ribonuclease Inhibitor. Our proprietary reaction buffer composition is optimized to increase specificity by delaying no-template control (NTC) amplification time.
Specifications
Master Mix | |
Bst DNA Polymerase | |
• 2X SuperScript IV RT-LAMP Master Mix (1.25 mL) |
|
65°C | |
Dry Ice | |
SYTO 9 Green Fluorescent Nucleic Acid Stain | |
5 to 15 minutes |
LAMP, RT-LAMP | |
1 x 1.25 mL | |
100 Reactions | |
SuperScript IV | |
Real-time and end-point detection | |
2X |