missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate

SuperSignal™ West Atto Ultimate Sensitivity Substrate is an enhanced chemiluminescent (ECL) substrate for high attogram protein detection and provides the high level sensitivity and better signal-to-noise ratios compared to other commercially available horseradish peroxidase (HRP) substrates.

Brand:  Thermo Scientific™ A38555

298.91 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 17181819

  • £421.00 / 100mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sensitivity: low-femtogram to high-attogram
  • Stability: eight hours for working solution, one year for kit at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: up to four hours
  • Recommended primary antibody concentration: 1:1000 to 1:5000 dilution (0.2 to 1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:100000 to 1:250000 dilution (4 to 10 ng/mL)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Substrate
Liquid
Nitrocellulose, PVDF
100 mL
Low-femtogram to high-attogram
SuperSignal West Atto Substrate / SuperSignal West Atto Stable Peroxide

• Substrate Solution, 50 mL
• Stable Peroxide Solution, 50 mL

Sufficient for 12 mini-gel blots; 1000 cm2 of membrane.

Store reagents at 2–8°C.
Chemiluminescence
Western Blot
SuperSignal™
Horseradish Peroxidase
12 Mini-Gel Blots, 1000 sq. cm of Membrane
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate > Quantity: 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.