Learn More
Thermo Scientific™ SuperSignal™ West Dura Extended Duration Substrate
Luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.
Brand: Thermo Scientific™ 34076
562.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : Weight : 0.20000kg
Description
Thermo Scientific™ SuperSignal West Dura Extended Duration Substrate is a luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.
SuperSignal West Dura Extended Duration Substrate for HRP is optimized for high sensitivity and long signal duration, making it ideal for cooled CCD camera-based detection systems and quantitation. Unlike substrates with signals that decline to barely detectable levels in 30–60 minutes, the signal produced with SuperSignal West Dura chemiluminescent substrate is stable for 24 hours, enabling multiple film or camera exposures.SuperSignal West Dura Extended Duration Substrate characteristics include:
- Sensitivity: mid-femtogram
- Stability: 24-hr working solution stability; 1-year kit stability at room temperature
- Compatibility: nitrocellulose and PVDF membranes
- Signal duration: 24 hours
- Recommended primary antibody concentration: 1:1,000–1:50,000 dilution (0.02–1.0 μg/mL)
- Recommended secondary antibody concentration: 1:50,000–1:250,000 dilution (4–20 ng/mL)
Specifications
Substrate | |
Liquid | |
Nitrocellulose, PVDF | |
200 mL | |
2°: 1:50-250K (4-20ng/mL), 1°: 1:5K (0.02- 1μg/mL) | |
24 hr. | |
Luminol/Enhancer, 100mL, Stable Peroxide Buffer, 100mL |
Chemiluminescent | |
Western Blot | |
SuperSignal™ | |
Horseradish Peroxidase | |
Mid-femtogram | |
25 mini-gel blots, 2000 sq. cm of membrane | |
Sufficient For: 25 mini-gel blots; 2000 cm2 of membrane • Luminol/Enhancer, 100 mL • Stable Peroxide Buffer, 100 mL Store at room temperature. |
For Research Use Only. Not for use in diagnostic procedures.