missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Dura Extended Duration Substrate

Luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.

Brand:  Thermo Scientific™ 37071

 View more versions of this product

Product Code. 10398773

  • £97.25 / 20mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ SuperSignal West Dura Extended Duration Substrate is a luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.

SuperSignal West Dura Extended Duration Substrate for HRP is optimized for high sensitivity and long signal duration, making it ideal for cooled CCD camera-based detection systems and quantitation. Unlike substrates with signals that decline to barely detectable levels in 30–60 minutes, the signal produced with SuperSignal West Dura chemiluminescent substrate is stable for 24 hours, enabling multiple film or camera exposures.

SuperSignal West Dura Extended Duration Substrate characteristics include:

  • Sensitivity: mid-femtogram
  • Stability: 24-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 24 hours
  • Recommended primary antibody concentration: 1:1,000–1:50,000 dilution (0.02–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:50,000–1:250,000 dilution (4–20 ng/mL)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Substrate
Liquid
Nitrocellulose, PVDF
20 mL
2°: 1:50-250K (4-20ng/mL), 1°: 1:5K (0.02- 1μg/mL)
24 hr.
Stable Peroxide Buffer, 10mL, Luminol/Enhancer, 10mL
Chemiluminescence
Western Blot
SuperSignal™
Horseradish Peroxidase
Mid-femtogram
2 Mini-Gel Blots, 200 sq. cm of Membrane
Sufficient For: 2 mini-gel blots; 200 cm2 of membrane
• Luminol/Enhancer, 10 mL
• Stable Peroxide Buffer, 10 mL

Store at room temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.