missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SuperSignal™ West Dura Extended Duration Substrate
Shop All Thermo Scientific Products
Click to view available options
Quantity:
20 mL
100 mL
200 mL
Unit Size:
100mL
200mL
20mL
Description
Thermo Scientific™ SuperSignal West Dura Extended Duration Substrate is a luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.
SuperSignal West Dura Extended Duration Substrate for HRP is optimized for high sensitivity and long signal duration, making it ideal for cooled CCD camera-based detection systems and quantitation. Unlike substrates with signals that decline to barely detectable levels in 30–60 minutes, the signal produced with SuperSignal West Dura chemiluminescent substrate is stable for 24 hours, enabling multiple film or camera exposures.
SuperSignal West Dura Extended Duration Substrate characteristics include:
- Sensitivity: mid-femtogram
- Stability: 24-hr working solution stability; 1-year kit stability at room temperature
- Compatibility: nitrocellulose and PVDF membranes
- Signal duration: 24 hours
- Recommended primary antibody concentration: 1:1,000–1:50,000 dilution (0.02–1.0 μg/mL)
- Recommended secondary antibody concentration: 1:50,000–1:250,000 dilution (4–20 ng/mL)
Specifications
Specifications
| Product Type | Substrate |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | SuperSignal™ |
| Quantity | 100 mL |
| Substrate | Horseradish Peroxidase |
| Recommended Antibody Concentrations | 2°: 1:50-250K (4-20ng/mL), 1°: 1:5K (0.02- 1μg/mL) |
| Sensitivity | Mid-femtogram |
| Show More |
Documents
Product Information
Manuals
User Guide: SuperSignal West Dura Extended Duration Substrate
Version: B.0
File size: 726.6 kB
Scientific Resources
Technical Notes
Expires: 2025-12-30
Protein Biology Essentials Promotion
Our top products empowering your research from start to finish
Expires: 2025-12-31
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
Expires: 2025-12-19
Seek Out Great Savings With the Lab of Curiosity
Every lab holds its own mysteries. What will you uncover? Step closer, search out your essentials, and discover how curiosity can lead to incredible savings. Promo code 24110.
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ SuperSignal™ West Dura Extended Duration Substrate >
Spot an opportunity for improvement?Share a Content Correction