missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Supor™ 200 Membrane Disc Filters

Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.

Brand:  Cytiva (formerly Pall Lab) 60300

94.53 GBP valid until 2024-12-31
Use promo code "21931" to get your promotional price.



Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 17154501

  • £108.00 / Pack of 100
Estimated Shipment: 30-08-2024
to see stock.
Add to basket

Description

Description

  • These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
  • For research use only
Specifications

Specifications

PES
Non-sterile
Plain
0.2 μm
145 μm
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers