missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Supor™ 200 Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
£103.00 - £677.00
Specifications
Material (Membrane) | PES |
---|---|
Pore Size | 0.2 μm |
Sterility | Non-sterile |
Thickness (Metric) | 145 μm |
Product Code | Brand | Diameter (Metric) Membrane | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Type | Price | Quantity & Availability | ||||
17154501
|
Cytiva (formerly Pall Lab)
60300 |
25 mm | Plain |
£108.00
Pack of 100 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
17164501
|
Cytiva (formerly Pall Lab)
60301 |
47 mm | Plain |
£319.92
Pack of 100 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
17144501
|
Cytiva (formerly Pall Lab)
60298 |
13 mm | Plain |
£103.00
Pack of 100 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
17174501
|
Cytiva (formerly Pall Lab)
60305 |
142 mm | Tabbed |
£260.00
Pack of 25 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
17184501
|
Cytiva (formerly Pall Lab)
60307 |
293 mm | Tabbed |
£550.00
Pack of 25 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
17144511
|
Cytiva (formerly Pall Lab)
60334 |
90 mm | Plain |
£677.00
Pack of 100 |
Estimated Shipment: 13-11-2024
Log in to see stock. |
||||
17154571
|
Cytiva (formerly Pall Lab)
66550 |
293 mm | Plain |
£404.00
Pack of 25 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
Description
- These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
- For research use only
Specifications
PES | |
0.2 μm | |
Non-sterile | |
145 μm |