Learn More
Cytiva (formerly Pall Lab) Supor™ 200 Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
Brand: Cytiva (formerly Pall Lab) 60301
276.76 GBP valid until 2024-12-31
Use promo code "21931" to get your promotional price.
Additional Details : Weight : 0.05700kg
Description
- These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
- For research use only
Specifications
PES | |
Non-sterile | |
Plain |
0.2 μm | |
145 μm | |
47 mm |