Learn More
Cytiva (formerly Pall Lab) Supor™ 800 Membrane Disc Filters
High flow rate membrane optimized for biological, pharmaceutical, and sterilizing filtration requirements.
Brand:  Cytiva (formerly Pall Lab) 66555
Additional Details : Weight : 0.29450kg
 View more versions of this product
Product Code. 17104581
  • £253.00 / Pack of 25
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
  • Fast filtration with superior flow rates and high throughputs
  • Low protein binding and extensive drug compatibility for critical applications
  • Saves time and money with fewer filter changes per sample volume
  • 142 and 293 mm discs feature printed tab for instant recognition of pore size and lot number
  • Designed for laboratory applications only
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
PES
Non-sterile
Plain
0.8 μm
140 μm
142 mm