Learn More
Invitrogen™ SureCast™ Gel Handcast Bundle A - Hardware and Reagents
Pour your own polyacrylamide gels easily and confidently with a SureCast™ Gel Handcast Bundle A. The SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.
Brand: Invitrogen™ HC1000SR
262.91 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes:
The SureCast Handcast Bundle A includes all the hardware and reagents you need to get started:
- 2 SureCast Handcast Stations
- 2 SureCast™ Sealing Pads
- 2 SureCast™ Glass Plate Sets (2 front and 2 back)
- 6 SureCast™ Gel Combs (2 each: 10-, 12-, 15-well)
- 10 SureCast™ Gel Spacers
- 1 SureCast™ Stacking Buffer (1 L), 2-pack
- 1 SureCast™ Resolving Buffer (1 L), 2-pack
- 1 SureCast™ Acrylamide Solution (40%) (450 mL)
- 1 SureCast™ APS (25 g)
- 1 SureCast™ TEMED (30 mL)
Description
Innovate, leak-free design—minimizes the number of failed gels, resulting in less wasted time
- Superior glass plate durability
- Simple assembly of casting components—just drop the glass plates in, then lock the cam handle
- Unique tilt feature for easy loading of acrylamide solutions
For a bundle that does not include the SureCast Handcast Reagents, please see SureCast™ Gel Handcast Bundle B. The system features durable glass plates that are securely sealed using a single-motion, load-and-lock mechanism. The unique tilt feature of the SureCast™ Handcast Station minimizes spillage and enhances loadability of casting solutions during handcasting. SureCast gels are designed for use in the Mini Gel Tank.

Specifications
• 2 SureCast Gel Handcast Stations • 2 sets SureCast Glass Plates • 10 SureCast Gel Spacers • 1 SureCast 10-well Multi-Use Tool • 1 SureCast 12-well Multi-Use Tool • 1 SureCast 15-well Multi-Use Tool • 1 set SureCast Gel Combs (10-, 12-, 15-well) • 2 x 500 mL SureCast Stacking Buffer packs. Store at room temperature. • 5 x 500 mL SureCast Stacking Buffer packs. Store at room temperature. • 2 x 500 mL SureCast Resolving Buffer packs. Store at room temperature. • 5 x 500 mL SureCast Resolving Buffer packs. Store at room temperature. • 25 g SureCast Ammonium Persulfate. Store at room temperature. • 450 mL SureCast Acrylamade Solution. Store at room temperature. • 30 mL SureCast TEMED. Store at room temperature. |
|
SureCast | |
1 kit |
Mini | |
Gel Handcast Bundle A | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.