Learn More
Fisherbrand™ SureOne™ Filter Tip Reload Pipette Tips
New, improved Fisherbrand SureOne Reload system offers a full line of micropoint universal fit pipet tips
£42.35 - £88.50
Specifications
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Refill
View More Specs
Products 4
Product Code Brand Volume (Metric) Tip Style Price Quantity & Availability  
Product Code Brand Volume (Metric) Tip Style Price Quantity & Availability  
11927724
View Documents Promotion Details
Fisherbrand™
0.1 to 20 μL Extended Length
£42.35
Pack of 960
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11967724
View Documents Promotion Details
Fisherbrand™
30 to 300 μL Extended Length
£88.50
Pack of 960
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11917724
View Documents Promotion Details
Fisherbrand™
0.1 to 10 μL Extended Length
£42.35
Pack of 960
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11977724
View Documents Promotion Details
Fisherbrand™
100 to 1000 μL Extended Length
£61.55
Pack of 560
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
Sterile and free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin and DNA
Packaging Features
  • Offset wells allow filter tips to stack
  • Rigid inserts are multi-channel format ensuring secure tip seal
  • Lot numbered inserts for traceability also include easy to read tip volume and Fisher catalog number
  • Sterile packaging with outer sleeve protects tips from contamination while refilling inserts
  • Environmentally friendly reload packaging reduces plastic waste and saves space
  • Blister sleeve guide rails align tip inserts for smooth and reliable transfer into the rack
Rack Features
  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Recycle Logo molded into all rack components
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable—all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact Clip helps prevent unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format
  • Robust non-pliable design provides secure seals on every tip
Specifications
Specifications
Filtered
Clear
Refill
Yes
Polypropylene
Sterile
Autoclavable
Extended Length
SureOne