Learn More
Thermo Scientific™ Surfact-Amps™ Detergent Sampler
Convenient collection of various Surfact-Amps Detergent Solutions and other selected useful detergents for protein research methods.
Brand: Thermo Scientific™ 28340
Code : PN
Additional Details : Weight : 0.10000kg Transport : UN number : 1481 Chem class : 5.1 Pack group : II
Description
Thermo Scientific™ Surfact-Amps Detergent Sampler is a convenient collection of various Thermo Scientific™ Surfact-Amps Detergent Solutions and selected other useful detergents for protein research methods.
The sampler kit is an economical way to test which detergent is most effective for a particular protein storage or experimental system. Thermo Scientific Surfact-Amps Purified Detergent Solutions are convenient 10% (w/v) solutions of commonly used detergents that provide unsurpassed purity, quality and stability for proteomics and molecular biology methods. Unlike neat detergent formulations, Surfact-Amps 10% Solutions are not so viscous that you can't aliquot them accurately. The detergent solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products. Don't begin your experiment with detergents of unknown age and purity! Just open an ampule and dilute the contents into your buffer at the desired concentration. Three of the ten detergents in the sampler kit are supplied as solids (100mg each) rather than 10% solutions.Highlights:
Surfact-Amps Detergent Solutions are
- Precise 10% detergent solutions in ultrapure water ampuled under nitrogen
- Easy to accurately dispense and dilute for use
- Exceptionally pure: less than 1.0 μeq/mL peroxides and carbonyls
- Highly stable: packed in glass ampules under inert nitrogen gas
Includes:
One ampule each of seven different detergent solutions and 100mg each of three additional detergent solids. Brij™, Triton™ and Tween™ are trademarks of their respective owners.
Specifications
Detergent | |
Liquid | |
Detergent Solution |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C. | |
Surfact-Amps™ | |
10 Vials |
For Research Use Only. Not for use in diagnostic procedures.