Learn More
SVL™ Sealing Ring for Butt Joint
Manufactured from silicone rubber
Brand: SVL 701-01
This item is not returnable.
View return policy
Description
- Silicone rubber and sheathed with PTFE for protection against chemical attack
- Internal diameter of ring corresponds to the bore of the SVL glass screwthread so that flow is not restricted
Specifications
Joint size: 15 | |
Butt joints | |
Silicone Rubber, PTFE (Sheath) | |
Sealing Ring |