Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Brand:  Thermo Scientific™ 60180-794
 View more versions of this product
Product Code. 10702327
Description
Description
Make column changeovers quick and easy. The Thermo Scientific™ SWAP-IT™ GC-MS Interface system is a powerful tool that enables capillary column changes without the need to vent your mass spectrometer (MS). It uses a specially deactivated, robust stainless steel transfer capillary as an interface between the analytical column and the MS detector. The small inner diameter of transfer capillary ensures minimum exposure to air and water when the analytical column is disconnected. A helium purge is not required. Finger-tight connections make it quick and easy without the need for tools.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GC-MS Interface Kit
SWAP-IT GC-MS Interface Kit
Interface, Nuts, Ferrules, and Accessories
Agilent GC-MS Instruments
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ SWAP-IT™ GC-MS Interface System > GCMS Interface Kit; For Agilent GC-MS Instruments