missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
Brand: Eppendorf™ 5702700002
Code : 99
Additional Details : Weight : 1.30000kg
Description
- Accommodates tubes and plates
- Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
- Rotor life time: max. 40,000 cycles
- Rotor, buckets, caps and adapters are autoclavable
Specifications
Swinging Bucket Rotor | |
4,400 rpm | |
8 x 15mL buckets |
2,800 x g | |
A-8-17 | |
8 |