missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
£290.00 - £2720.00
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
10258211
|
Eppendorf™
5702720003 |
A-4-38 | 4 x 85mL buckets |
£1025.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10054271
|
Eppendorf™
5804719000 |
A-4-44 | 4 hangers for 2 x 50mL Falcon-G™ |
£1620.00
Each |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10794563
|
Eppendorf™
5810725003 |
A-4-81-MTP | 4 MTP buckets |
£2720.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10120052
|
Eppendorf™
5810718007 |
A-4-81 | 4 x 500mL rectangular buckets |
£2720.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10282811
|
Eppendorf™
5804709004 |
A-4-44 | 4 x 100mL rectangular buckets |
£1345.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10084161
|
Eppendorf™
5702700002 |
A-8-17 | 8 x 15mL buckets |
£290.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10580152
|
Eppendorf™
5810709008 |
A-4-62 | 4 x 250mL rectangular buckets |
£1855.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10394792
|
Eppendorf™
5810711002 |
A-4-62-MTP | 4 MTP buckets |
£1775.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Accommodates tubes and plates
- Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
- Rotor life time: max. 40,000 cycles
- Rotor, buckets, caps and adapters are autoclavable
Specifications
Swinging Bucket Rotor |