missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
Brand: Eppendorf™ 5702720003
Code : 99
Additional Details : Weight : 2.23560kg
Description
- Accommodates tubes and plates
- Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
- Rotor life time: max. 40,000 cycles
- Rotor, buckets, caps and adapters are autoclavable
Specifications
Swinging Bucket Rotor | |
4,400 rpm | |
4 x 85mL buckets |
3,000 x g | |
A-4-38 | |
4 |