Learn More
OHAUS™ Swivel Multi-Purpose Clamps
OHAUS™ Swivel Multi-Purpose Clamps are designed to securely grip and position a variety of laboratory glassware.
Brand:  OHAUS™ 30392226
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15702279
  • £92.50 / Each
Estimated Shipment: 11-10-2024
to see stock.
Add to basket
Description
Description
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
  • Available as single or dual-adjust in nickel-plated zinc or stainless steel
  • Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
Specifications
Specifications
Multi-purpose Clamp
Non-slip vinyl sleeves and fiberglass covers
6.42 in.
Medium
2-Prong Single Adjust
Stainless Steel
Clamp, Multi Purpose, CLM-SWIVL2SSM
163.00 mm
0 to 76 mm