missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Symmetry C8 Column
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactivated silica.
Brand: Waters Corporation 186002609

This item is not returnable.
View return policy
Description
- Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
Specifications
2.1 mm | |
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
Yes |
100 mm | |
Spherical | |
100 Å | |
L7 | |
Column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation Symmetry C8 Column
Spot an opportunity for improvement?Share a Content Correction