missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Symmetry C8 Prep Column
Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.
Brand: Waters Corporation 186000229
This item is not returnable.
View return policy
Description
- No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
- Available in a variety of particle sizes, chemistries and dimensions.
Specifications
19 mm | |
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
Yes |
100 mm | |
Spherical | |
100 Å | |
L7 | |
Preparative Column |