missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Symmetry C8 Prep Column
Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.
£892.50 - £7410.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 100 Å |
Quantity | 1/Ea. |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
13200150
|
Waters Corporation
186000214 |
50 mm | 7.8 mm |
£892.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
||||
13210150
|
Waters Corporation
186000215 |
100 mm | 7.8 mm |
£1222.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
||||
13260150
|
Waters Corporation
186000229 |
100 mm | 19 mm |
£3555.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
||||
13200160
|
Waters Corporation
186000237 |
50 mm | 30 mm |
£5205.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
||||
13210160
|
Waters Corporation
186000238 |
100 mm | 30 mm |
£7410.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
||||
Description
- No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
- Available in a variety of particle sizes, chemistries and dimensions.
Specifications
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
Yes |
Spherical | |
100 Å | |
L7 | |
Preparative Column |