Learn More
Agilent BioTek Synergy LX Multi-Mode Reader
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
The BioTek Synergy LX multi-mode reader automates many common microplate assays. For Research Use Only. Not for use in diagnostic procedures.
Brand: Agilent BioTek SLXATS-SN
Description
The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability
The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.
Certifications
CE and TUV marked. RoHS compliant.
Services
12 Months
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
200 to 900 nm | |
CE and TUV marked. RoHS compliant. | |
38.1 cm | |
24 sec. (96 wells), 76 sec. (384 wells) | |
Monochromator | |
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability | |
50/60 Hz | |
<1% from 0 to 2.5 OD | |
SLXATS | |
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) | |
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP | |
English | |
100/240 V | |
12.3 kg | |
38.1 cm | |
Xenon Flash |
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD | |
≤5 nm | |
15 in. | |
Linear, Orbital, Double Orbital | |
Monochromator Bandpass | |
15 in. | |
38.1 cm | |
Absorbance; Touchscreen | |
Multi-mode Microplate Reader | |
0.001 OD | |
2 | |
1 year | |
27 lb. | |
15 in. | |
Photodiode, PMT | |
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.