Learn More
Corning™ Synthemax™ II Dissolvable Microcarriers
Corning microcarriers are designed to simplify your scale-up processes.
Brand:  Corning™ 7288
 View more versions of this product
Product Code. 17233084
  • £152.00 / 1g
Explore more special offers
Description
Description
  • Sterile
  • Ready-to-use
  • Available in closed systems packaging to be used directly with bioreactors
  • Additionally, the microcarriers are surface-treated to enhance cell attachment, maximizing cell yield and viability
  • Consistency – USP Class VI polystyrene material provides a consistent platform
  • Performance – Corning microcarriers are offered untreated or with the following surface treatments
  • Corning Synthemax™ II substrate creates a synthetic surface on the microcarriers for stem cell expansion
  • For use with MSC and iPS/ES applications. (Animal Origin Free)
  • Sterility Assurance Level (SAL) of 10−6
  • Nonpyrogenic
  • Available as closed system solutions
  • Corning microcarriers are available sterile and ready to use in 1 to 10 g vials. This format provides flexibility to select the amount of beads to use.
  • During the mature phase of process development and into manufacturing, closed system packaging would be necessary to maintain sterility and ease bioreactor loading.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSC and iPS/ES Applications
Synthemax™ II
USP Class VI Polystyrene
Sterile
Microcarrier
1 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Synthemax™ II Dissolvable Microcarriers > 1 g