Learn More
BRAND™ T-Shape Tubing Connector
Manufactured from translucent polypropylene, Brand™ T-Shape Tubing Connectors can be repeatedly autoclaved at 121°C.
Brand:  BRAND™ 152815
24.75 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11823930
  • £32.35 / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Good chemical resistance and mechanical properties
  • Straight-through tube and side arm
  • Provided in packs of 20 (152815) and 10 (152825)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Length of straight-through tube: 69mm; length of side arm including diameter of straight-through tube: 47mm
Polypropylene
20/Pk.
Autoclavable
Tubing with inner diameter of 10-11mm
Translucent